missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate Anhydrous, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10345720
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

HNa2O4P, CAS Number-7558-79-4, disodium orthophosphate, acetest, disodium hydrogen phosphate, disodium hydrogenorthophosphate, disodium phosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt, soda phosphate, sodium phosphate dibasic, dibasic sodium phosphate, 3kg, CHEBI:34683, White

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
pH 8.7 to 9.3
Physical Form Powder/Solid
Quantity 500 g
IUPAC Name disodium hydrogen phosphate
Formula Weight 141.96g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.