missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate Dihydrate, HPLC for Electrochemical Detection, Certified, Fisher Chemical

Product Code. 10776491
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 243°C
CAS 10028-24-7
Color White
pH 8.7 to 9.4
Physical Form Solid
Quantity 500 g
Molecular Formula H5Na2O6P
MDL Number MFCD00149182
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium;hydrogen phosphate;dihydrate
Molecular Weight (g/mol) 177.99
PubChem CID 6451167
Formula Weight 177.99g/mol
Packaging Plastic powder jar
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'