missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

58.39 € - 369.84 €

Specifications

Chemical Name or Material Di-tert-butyl dicarbonate
Name Note 1M Solution in THF
CAS 24424-99-5, 109-99-9
Color Colorless
Density 0.9130g/mL
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10400704
View Documents
Thermo Scientific Acros
426801000
100 mL AcroSeal™ Glass Bottle
58.39 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10525524
View Documents
Thermo Scientific Acros
426808000
800 mL AcroSeal™ Glass Bottle
369.84 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Di-tert-butyl dicarbonate
24424-99-5, 109-99-9
0.9130g/mL
AcroSeal™ Glass Bottle
O[CO2C(CH3)3]2
15, 3038
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
218.25
CHEBI:48500
Liquid
1M Solution in THF
Colorless
−17°C
C10H18O5
MFCD00008805
0.913
DYHSDKLCOJIUFX-UHFFFAOYSA-N
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
90495
218.25
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Fatal if inhaled.
May cause drowsiness

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 246-240-1

Product Identifier
  • Di-tert-butyl dicarbonate, 1M solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.