missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-tert-butyl malonate, 98%, Thermo Scientific Chemicals
51.80 € - 175.00 €
Chemical Identifiers
CAS | 541-16-2 |
---|---|
MDL Number | MFCD00008810 |
InChI Key | CLPHAYNBNTVRDI-UHFFFAOYSA-N |
Synonym | di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate |
PubChem CID | 68324 |
IUPAC Name | ditert-butyl propanedioate |
SMILES | CC(C)(C)OC(=O)CC(=O)OC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10700482
|
Thermo Scientific Acros
406620100 |
10 g |
51.80 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10764204
|
Thermo Scientific Acros
406620500 |
50 g |
175.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
541-16-2 | |
CLPHAYNBNTVRDI-UHFFFAOYSA-N | |
68324 | |
CC(C)(C)OC(=O)CC(=O)OC(C)(C)C |
MFCD00008810 | |
di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate | |
ditert-butyl propanedioate |
Specifications
541-16-2 | |
Colorless to Yellow | |
110°C to 111°C (22.0 mmHg) | |
Authentic | |
Glass bottle | |
CH2(CO2C(CH3)3)2 | |
MFCD00008810 | |
01,210 | |
0.966 | |
CLPHAYNBNTVRDI-UHFFFAOYSA-N | |
ditert-butyl propanedioate | |
68324 | |
98% | |
Di-tert-butyl malonate |
-7°C | |
0.9660g/mL | |
88°C | |
97.5% min. (GC) | |
C11H20O4 | |
1.4175 to 1.4195 | |
02, III, 1621 | |
15, 3041 | |
di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate | |
CC(C)(C)OC(=O)CC(=O)OC(C)(C)C | |
216.28 | |
216.28 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 208-769-6
Product Identifier
- Di-tert-butyl malonate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title