missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl peroxide, 99%

Product Code. 10607963 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Plastic bottle
Unit Size:
100g
250g
5g
This item is not returnable. View return policy

Product Code. 10607963

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-05-4 | C8H18O2 | 146.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-05-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00008803
InChI Key LSXWFXONGKSEMY-UHFFFAOYSA-N
Synonym di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d
PubChem CID 8033
SMILES CC(C)(C)OOC(C)(C)C

Specifications

Melting Point -30°C
Density 0.8000g/mL
Boiling Point 109°C to 110°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 0.1% max. Tert-butyl hydroperoxide (GC)
Packaging Plastic bottle
Linear Formula (CH3)3COOC(CH3)3
Refractive Index 1.3880 to 1.39
Quantity 250 g
Merck Index 15, 3508
Specific Gravity 0.8
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in most organic solvents
IUPAC Name 2-tert-butylperoxy-2-methylpropane
Viscosity 0.9 mPa.s (20°C)
Formula Weight 146.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Di-tert-butyl peroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Di-tert-butyl peroxide
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Organic peroxide Type E & F
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H412-Harmful to aquatic life with long lasting effects.
  • H242-Heating may cause a fire.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.