missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Diacetone-D-glucose, 98+% Product Code.: 10182280

Thermo Scientific Chemicals Diacetone-D-glucose, 98+%

Product Code. 10182280
100 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10182280

Brand: Thermo Scientific Chemicals 112011000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 582-52-5
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00005544
InChI Key KEJGAYKWRDILTF-MQIGXGKASA-N
Synonym 1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-olShow MoreShow Less
PubChem CID 40469256
IUPAC Name (3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

Specifications

Melting Point 107.0°C to 110.0°C
Color White to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 31,155
Merck Index 15,2968
Specific Rotation Condition − 11.00 (20.00°C c=5,EtoH)
Specific Rotation − 11.00
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in chloroform,alcohol,acetone,and warm ether
Formula Weight 260.28
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material Diacetone-D-glucose, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0410800 Certificate TypeCertificate of Analysis Date26/02/2024 Catalog Number112015000,112010250,112011000
Lot NumberA0456200 Certificate TypeCertificate of Analysis Date24/11/2023 Catalog Number112015000,112010250,112011000
Lot NumberA0421795 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number112015000,112010250,112011000
Lot NumberA0395251 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number112015000,112010250,112011000
Lot NumberA0429158 Certificate TypeCertificate of Analysis Date28/04/2021 Catalog Number112015000,112010250,112011000
Product Identifier
  • Diacetone-D-glucose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Diacetone-D-glucose, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.