Learn More
MP Biomedicals Diagnostic Kit for Cardiac Troponin I (MPQuanti™ only)
Diagnostic Kit for Cardiac Troponin I is intended for in-vitro quantitative determination of human cardiac Troponin I (cTnI) in whole blood, serum, or plasma.
Brand:  MP Biomedicals 07CTN15025
Product Code. 30280598
  • 52.25 € / 1 set
Explore more special offers
Description
Description
  • Diagnostic Kit for Cardiac Troponin I (Whole Blood/Serum/Plasma) is based on fluorescence immunoassay technology.
  • The sample moves through the strip from sample pad to absorbent pad.
  • If the specimen contains cTnI, it attaches to the fluorescent microspheres-conjugated anti-cTnI antibodies.
  • Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
  • The concentration of cTnI in the sample correlates with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and product standard curve, the concentration of cTnI in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in vitro diagnostic use
  • Working with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 1 ̴40 ng/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
  • Used as an aid in diagnosis of Myocardial Infarction (MI)
Specifications
Specifications
Troponin I
Human whole blood, Human serum, Human plasma
POC, cTnI, Quantitative
Ready-to-Use (RTU)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals Diagnostic Kit for Cardiac Troponin I (MPQuanti™ only) > Cardiac Troponin 1