Learn More
MP Biomedicals Diagnostic Kit for D-Dimer (MPQuanti™ only)
Diagnostic Kit for D-Dimer is intended for in-vitro quantitative detection of D-Dimer in human whole blood or plasma.
Brand:  MP Biomedicals 07CTNL5025
Product Code. 30280622
  • 65.32 € / 1 set
Explore more special offers
Description
Description
  • Diagnostic Kit for D-Dimer (Whole Blood/Plasma) is based on fluorescence immunoassay technology to measure D-Dimer in human whole blood or plasma as an aid in the diagnosis of DVT and PE.
  • The test is intended for healthcare professionals use.
  • The sample moves through the strip from sample pad to absorbent pad. If the specimen contains D-Dimer, it attaches to the fluorescent microspheres-conjugated anti-D-Dimer antibodies.
  • Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
  • The concentration of D-Dimer in the sample correlates with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and the standard curve, the concentration of D-Dimer in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 1 ̴10 mg/L, R≥0.990
  • Test results should be interpreted at 15 minutes
  • Used as an aid in the diagnosis of DVT and PE
Specifications
Specifications
D-dimer
Human whole blood, Human plasma
POC, Quantitative, D-Dimer
Ready-to-Use (RTU)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals Diagnostic Kit for D-Dimer (MPQuanti™ only) > D-Dimer