Learn More
MP Biomedicals Diagnostic Kit for Myoglobin (MYO) (MPQuanti™ only)
Diagnostic Kit for Myoglobin (MYO), is intended for in-vitro quantitative determination of Myoglobin (MYO) in human whole blood, serum, or plasma.
Brand:  MP Biomedicals 07MY5025
Product Code. 30280608
  • 52.25 € / 1 set
Explore more special offers
Description
Description
  • Diagnostic Kit for Myoglobin (MYO) (Whole blood/serum/plamsa) is based on fluorescence immunoassay technology.
  • The sample moves through the strip from sample pad to absorbent pad.
  • If the specimen contains Myoglobin, it attaches to the fluorescent microspheres-conjugated anti-Myoglobin antibodies.
  • Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
  • The concentration of Myoglobin in the sample correlates with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and the standard curve, the concentration of Myoglobin in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 5 ̴ 200 ng/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
  • For the diagnosis of myocardial infarction (MI)
Specifications
Specifications
Myoglobin
Human whole blood, Human serum, Human plasma
Ready-to-Use (RTU)
Immunofluorescence
POC, Quantitative, Myoglobin, Cardiac
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals Diagnostic Kit for Myoglobin (MYO) (MPQuanti™ only) > MYO