Learn More
MP Biomedicals Diagnostic Kit for Progesterone (MPQuanti™ only)
Diagnostic Kit for Progesterone is intended for in-vitro quantitative determination of Progesterone in human serum, and plasma.
Brand:  MP Biomedicals 07PROG5025
Product Code. 30280621
  • 97.97 € / 1 set
Explore more special offers
Description
Description
  • Diagnostic Kit for Progesterone (Serum/Plasma) is based on fluorescence immunoassay technology.
  • The specimen moves through the strip from sample pad to absorbent pad. Progesterone in the specimen will compete with the Progesterone antigen coated on the membrane with the progesterone antibody labeled with fluorescent microspheres.
  • The less Progesterone in the specimen, the more chance that fluorescent microspheres-conjugated anti-Progesterone antibodies can be captured by the Progesterone antigen coated on the membrane (Test line).
  • The concentration of Progesterone in the specimen is inversely related to the intensity of the fluorescent signal captured on the T line.
  • According to the fluorescence intensity of the test and the standard curve, the concentration of Progesterone in the specimen is calculated by the MPQuanti™ Immuflorescence Analyzer to show Progesterone concentration in the sample.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 1.4 ̴ 60 ng/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
Specifications
Specifications
Progesterone
Human serum, Human plasma
Ready-to-Use (RTU)
Immunofluorescence
POC, Quantitative, progesterone, hormone
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals Diagnostic Kit for Progesterone (MPQuanti™ only) > Progesterone