Learn More
VEE GEE Dial Thermometer
Dial thermometers of varying sizes based on personal preferences.
Brand:  VEE GEE 82160-6
 View more versions of this product
Product Code. 17821204
  • 27.20 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The dial thermometer shows the temperature using a metal stem and a circular scale.
  • It doesn't need batteries, it's tough, and you can change the readings by using the calibration screw on the back.
Specifications
Specifications
7.6 cm Diameter
1/2 in. NPT male thread on the back for threading into a pipeline and an adjustment screw for field calibration.
15.2 cm
Serialized Glass Thermometer
160°F
-40°F
-40 to +160°F
±1 subdivision
15.2 cm
Food & beverage quality control, general lab use, and asphalt/concrete testing
12 in.
Metal
Case is hermetically sealed to prevent lens fogging
71.1°C
-40°C
-40 to +71.1°C
2°F