Learn More
VEE GEE Dial Thermometer
Dial thermometers of varying sizes based on personal preferences.
Brand: VEE GEE 82160-12
This item is not returnable.
View return policy
Description
- The dial thermometer shows the temperature using a metal stem and a circular scale.
- It doesn't need batteries, it's tough, and you can change the readings by using the calibration screw on the back.
Specifications
7.6 cm Diameter | |
1/2 in. NPT male thread on the back for threading into a pipeline and an adjustment screw for field calibration. | |
30.5 cm | |
Serialized Glass Thermometer | |
160°F | |
-40°F | |
-40 to +160°F | |
±1 subdivision | |
30.5 cm |
Food & beverage quality control, general lab use, and asphalt/concrete testing | |
6 in. | |
Metal | |
Case is hermetically sealed to prevent lens fogging | |
71.1°C | |
-40°C | |
-40 to +71.1°C | |
2°F |