Learn More
Scotch-Brite™ Diamond Floor Pads, Purple
Diamond-coated floor pads for cleaning and polishing marble, terrazzo, slate, and polished concrete flooring. Helps eliminate floor finish and reduce labor costs.
Brand:  Scotch-Brite™ 7100159505
 View more versions of this product
Product Code. 17118800
  • 550.00 € / Pack of 5
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Simultaneously cleans and polishes flooring to achieve an outstanding shine
  • Ideal for marble, terrazzo, vinyl, slate, and polished concrete floors
  • Easy to use: no expensive or harmful chemicals needed, only water
  • Recommended for use with low and standard speed machines
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cleaning Pads
685 mm
Marble, Terrazzo, Slate, and Polished Concrete Flooring
Round
Purple
Burnishing, Cleaning
Diamond-coated