Learn More
Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Diamond powder is generally used as an abrasive to grind and polish hard surfaces
128.64 € - 454.26 €
Specifications
Chemical Name or Material | Diamond powder |
---|---|
Name Note | Synthetic |
CAS | 7782-40-3 |
Physical Form | Powder |
Assay Percent Notes | (metals basis) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366718
|
Thermo Scientific Alfa Aesar
013402.03 |
1 g |
128.64 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376718
|
Thermo Scientific Alfa Aesar
013402.06 |
5 g |
454.26 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Diamond powder is generally used as an abrasive to grind and polish hard surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diamond powder | |
7782-40-3 | |
(metals basis) | |
99.9% |
Synthetic | |
Powder | |
MFCD00211867 |
Safety and Handling
missing translation for 'einecsNumber' : 231-953-2
missing translation for 'rtecsNumber' : HL4158550
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.