missing translation for 'onlineSavingsMsg'
Learn More

Notrax Diamond Sof-Tred™ Dyna-Shield™ Layer with Non-Directional Deck Plate Pattern

Product Code. p-7227310
Click to view available options
Dimensions (L x W):
91 x 60 cm
150 x 91 cm
1830 x 60 cm
1830 x 91 cm
1830 x 122 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17772118

Brand: Notrax 419S0023BL

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Notrax Diamond Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.

  • Non-directional ''Deck Plate'' pattern allows freedom of movement
  • Exceptional worker comfort due to 127 mm thick specially formulated compound
  • Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
  • Available with yellow borders along sides comply with OSHA code 1910-144
  • All 4 sides are bevelled and sealed to minimize tripping hazards

Specifications

Dimensions (L x W) 91 x 60 cm
Color Black
Length (English) 36 in.
Length (Metric) 91 cm
Material Microcellular Vinyl
Type Industrial and Commercial Matting
Width (Metric) 60 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.