missing translation for 'onlineSavingsMsg'
Learn More

Notrax Diamond Stat™ Static Dissipative Anti-Fatigue Mat

Ergonomic benefit derived from 14 mm thick worker platform of a static dissipative combination of tough top surface laminated onto a comfortable sponge base for maximum durability coupled with fatigue relief.

Brand:  Notrax 826C0036BL

 View more versions of this product

Product Code. 17762278

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ergonomic benefit derived from 14 mm thick worker platform of a static dissipative combination of tough top surface laminated onto a comfortable sponge base for maximum durability coupled with fatigue relief
  • Equipped with 10 mm female socket attached to the mat and grounding cord
  • Diamond deck plate surface provides traction and allows easy twist turns
  • Featured with RedStop™ slip-resistant backing to prevent mats from shifting
  • UniFusion™ technology engineered to resist in the harshest work environments
  • Static dissipative anti-fatigue mats protect against static shocks while providing comfort for standing workers derived from a thick worker platform with vinyl sponge base for fatigue relief
  • Bevelled edges on all four sides to minimize risk of tripping
Specifications

Specifications

Black
2286 cm
Industrial and Commercial Matting
75 ft.
Conductive PVC
91 cm
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales