missing translation for 'onlineSavingsMsg'
Learn More

Diatomaceous Earth, Flux-calcined, Hyflo™ Super Cel™, Honeywell Fluka™

Product Code. 15605860
Click to view available options
Quantity:
5 kg
Packaging:
Fiber drum
This item is not returnable. View return policy
This item is not returnable. View return policy

Filter aid, flux calcined, treated with sodium carbonate

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 kg
CAS 68855-54-9
Packaging Fiber drum
Molecular Formula CNa2O3
MDL Number MFCD00132803
UN Number NONH for all modes of transport
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES C(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;carbonate
Molecular Weight (g/mol) 105.988
PubChem CID 10340
ChEBI CHEBI:29377
Chemical Name or Material Diatomaceous Earth, Flux-Calcined
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.