missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzothiophene-4-boronic acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 285.0°C to 290.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Quantity | 5 g |
IUPAC Name | dibenzothiophen-4-ylboronic acid |
Formula Weight | 228.08 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | Dibenzothiophene-4-boronic acid |
Safety and Handling

Product Identifier
- Dibenzothiophene-4-boronic acid
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dibenzothiophene-4-boronic acid, 95%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction