Learn More
(-)-Dibenzoyl-L-tartaric acid hydrate, 98%, Thermo Scientific Chemicals
35.40 € - 414.00 €
Chemical Identifiers
CAS | 62708-56-9 |
---|---|
Molecular Formula | C18H14O8 |
Molecular Weight (g/mol) | 358.30 |
MDL Number | MFCD00149119 |
InChI Key | YONLFQNRGZXBBF-OKILXGFUSA-N |
Synonym | 2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152 |
PubChem CID | 44119738 |
SMILES | OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483247
|
Thermo Scientific Alfa Aesar
A16180.22 |
100 g |
35.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493247
|
Thermo Scientific Alfa Aesar
A16180.36 |
500 g |
117.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403257
|
Thermo Scientific Alfa Aesar
A16180.0E |
2500 g |
414.00 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.
Solubility
Soluble in methanol, acetone and organic solvents. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
62708-56-9 | |
358.30 | |
YONLFQNRGZXBBF-OKILXGFUSA-N | |
44119738 |
C18H14O8 | |
MFCD00149119 | |
2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152 | |
OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O |
Specifications
62708-56-9 | |
C18H14O8 | |
2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152 | |
YONLFQNRGZXBBF-OKILXGFUSA-N | |
(2R,3S)-2,3-dibenzoyloxybutanedioic acid;hydrate | |
44119738 | |
≥98% |
90°C to 92°C | |
MFCD00149119 | |
Soluble in methanol,acetone and organic solvents. Slightly soluble in water. | |
OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O | |
358.30 | |
376.33 (358.31 Anhydrous) | |
(-)-Dibenzoyl-L-tartaric acid monohydrate |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
missing translation for 'einecsNumber' : 613-072-9
missing translation for 'rtecsNumber' : WW8070000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only