missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl phosphite, 95%, Thermo Scientific Chemicals

Product Code. 10135583
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17176-77-1
Molecular Formula C14H14O3P
Molecular Weight (g/mol) 261.24
MDL Number MFCD00004774
InChI Key RQKYHDHLEMEVDR-UHFFFAOYSA-N
Synonym dibenzyl phosphonate, phosphonic acid dibenzyl ester, unii-1o720l5h5a, phosphonic acid, bis phenylmethyl ester, dibenzylphosphit, dibenzylphosphonate, phosphonic acid bis phenylmethyl ester, oxo-bis phenylmethoxy phosphanium, mibxhgzaarwagi-uhfffaoysa-n, dibenzyl phosphite, technical grade
PubChem CID 6334615
SMILES O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.1800g/mL
Boiling Point 110.0°C to 120.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5CH2O)2P(O)H
Refractive Index 1.5500 to 1.5560
Quantity 5 g
Fieser 01,198
Merck Index 15,3019
Specific Gravity 1.18
IUPAC Name bis(benzyloxy)(oxo)-λ⁵-phosphanylium
Formula Weight 262.23
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Dibenzyl phosphite, tech., 80%
Show More Show Less
compliance-icons
Product Identifier
  • Dibenzyl phosphite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dibenzyl phosphite, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.