missing translation for 'onlineSavingsMsg'
Learn More
Dibenzyl phosphite, 95%, Thermo Scientific Chemicals Product Code.: 10162710

Dibenzyl phosphite, 95%, Thermo Scientific Chemicals

Product Code. 10162710
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Questo articolo non è restituibile. Consulta la politica di reso

Product Code. 10162710

Brand: Thermo Scientific Acros 112660250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 17176-77-1
Molecular Formula C14H14O3P
Molecular Weight (g/mol) 261.24
MDL Number MFCD00004774
InChI Key RQKYHDHLEMEVDR-UHFFFAOYSA-N
Synonym dibenzyl phosphonate, phosphonic acid dibenzyl ester, unii-1o720l5h5a, phosphonic acid, bis phenylmethyl ester, dibenzylphosphit, dibenzylphosphonate, phosphonic acid bis phenylmethyl ester, oxo-bis phenylmethoxy phosphanium, mibxhgzaarwagi-uhfffaoysa-n, dibenzyl phosphite, technical gradeShow MoreShow Less
PubChem CID 6334615
SMILES O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifica

Color Colorless to Yellow
Density 1.1800g/mL
Boiling Point 110.0°C to 120.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5CH2O)2P(O)H
Refractive Index 1.5500 to 1.5560
Quantity 25 g
Fieser 01,198
Merck Index 15,3019
Specific Gravity 1.18
IUPAC Name bis(benzyloxy)(oxo)-λ⁵-phosphanylium
Formula Weight 262.23
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Dibenzyl phosphite, tech., 80%
Vedi altri risultati Mostra meno risultati

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0449032 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number112660250,112661000,112660050
Lot NumberA0409503 Certificate TypeCertificate of Analysis Date18/04/2022 Catalog Number112660250,112661000,112660050
Lot NumberA0438431 Certificate TypeCertificate of Analysis Date25/01/2022 Catalog Number112660250,112661000,112660050
compliance-icons
Identificativo prodotto
  • Dibenzyl phosphite
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Dibenzyl phosphite, 95%, Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato