missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl phthalate, 97%

Product Code. p-7044172
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11361787

Brand: Thermo Scientific Alfa Aesar L04985.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 523-31-9 | C22H18O4 | 346.382 g/mol

Dibenzyl phthalate is an plasticizer used in toys that was found to exhibit potential estrogen agonist/ antagonist properties. It is used in PVC, paints, acrylic coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl phthalate is an plasticizer used in toys that was found to exhibit potential estrogen agonist/ antagonist properties. It is used in PVC, paints, acrylic coatings.

Solubility
Insoluble in water.

Notes
Incompatible materials are reducing agents. Strong acids. Strong bases and Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 523-31-9
Molecular Formula C22H18O4
Molecular Weight (g/mol) 346.382
MDL Number MFCD00014437
InChI Key UCVPKAZCQPRWAY-UHFFFAOYSA-N
Synonym dibenzyl phthalate, benzyl phthalate, 1,2-benzenedicarboxylic acid, bis phenylmethyl ester, phthalic acid, dibenzyl ester, dibenzylphthalate, 1, bis phenylmethyl ester, acmc-1b21b, phthalic acid dibenzyl ester, phthalic acid, bis-benzylester, dibenzyl phthalate, analytical standard
PubChem CID 220773
IUPAC Name dibenzyl benzene-1,2-dicarboxylate
SMILES C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2C(=O)OCC3=CC=CC=C3

Specifications

Melting Point 40°C to 42°C
Boiling Point 276°C to 278°C (15 mmHg)
Quantity 5 g
Beilstein 2338250
Solubility Information Insoluble in water.
Formula Weight 346.39
Percent Purity 97%
Chemical Name or Material Dibenzyl phthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.