missing translation for 'onlineSavingsMsg'
Learn More

Dibromomethane, 99%

Product Code. 10615532 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10615532

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74-95-3 | CH2Br2 | 173.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-95-3
Molecular Formula CH2Br2
Molecular Weight (g/mol) 173.83
MDL Number MFCD00000168
InChI Key FJBFPHVGVWTDIP-UHFFFAOYSA-N
Synonym methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068
PubChem CID 3024
ChEBI CHEBI:47077
IUPAC Name dibromomethane
SMILES C(Br)Br

Specifications

Melting Point -52.0°C
Color Colorless to Brown
Density 2.4700g/mL
Boiling Point 96.0°C to 98.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2Br2
Refractive Index 1.5400 to 1.5420
Quantity 100 mL
Beilstein 01,67
Fieser 11,337; 12,322; 16,393
Merck Index 15,6133
Specific Gravity 2.47
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol, ether, acetone and, chloroform
Formula Weight 173.83
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dibromomethane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Dibromomethane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.