missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin dichloride, 97%, Thermo Scientific Chemicals

Product Code. 10581184
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 683-18-1
MDL Number MFCD00000518
InChI Key RJGHQTVXGKYATR-UHFFFAOYSA-L
Synonym dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride
PubChem CID 12688
IUPAC Name dibutyl(dichloro)stannane
SMILES CCCC[Sn](CCCC)(Cl)Cl

Specifications

Melting Point 39.0°C to 41.0°C
Color White to Beige
Density 1.4000g/mL
Boiling Point 135.0°C (10.0 mmHg)
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass bottle
Molecular Formula C8H18Cl2Sn
Linear Formula [CH3(CH2)3]2SnCl2
Quantity 25 g
Fieser 01,213; 01,1294; 05,622
Specific Gravity 1.4
Solubility Information Solubility in water: 320mg/l,hydrolises in hot water. Other solubilities: soluble in many organic solvents
Molecular Weight (g/mol) 303.83
Formula Weight 303.83
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Dibutyltin dichloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dibutyltin dichloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'