missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(1,5-cyclooctadiene)platinum(II), 98%, Thermo Scientific Chemicals

Product Code. 10540934
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12080-32-9
Molecular Formula C8H12Cl2Pt
Molecular Weight (g/mol) 374.17
MDL Number MFCD00012413
InChI Key VVAOPCKKNIUEEU-PHFPKPIQSA-L
Synonym dichloro 1,5-cyclooctadiene platinum ii, 1,5-cyclooctadienedichloroplatinum, dichloro 1,5-cyclooctadiene platinum, platinum, dichloro 1,5-cyclooctadiene, 1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride, platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, 1,5-cyclooctadieneplatinum ii dichloride, dichloro 1,5-cyclooctadiene platinum ii 250mg, dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum, platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
PubChem CID 6436378
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride
SMILES Cl[Pt]Cl.C1C\C=C/CC\C=C/1

Specifications

Melting Point 285.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Pt)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble in water
Formula Weight 374.18
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Dichloro(1, 5-cyclooctadiene)platinum(II)
Show More Show Less
compliance-icons
Product Identifier
  • Dichloro(1,5-cyclooctadiene)platinum(II)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'