missing translation for 'onlineSavingsMsg'
Learn More
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), Pd 12.1%, Thermo Scientific Chemicals Product Code.: 15465025

Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), Pd 12.1%, Thermo Scientific Chemicals

Product Code. 15465025
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15465025

Brand: Thermo Scientific Alfa Aesar 046665.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205319-10-4
Molecular Formula C39H32Cl2OP2Pd
Molecular Weight (g/mol) 755.95
MDL Number MFCD14155707
InChI Key MNBAPNBMBOYNAW-UHFFFAOYSA-L
Synonym dichloro 9,9-dimethyl-4,5-bis diphenylphosphino xanthene palladium ii, palladium 2+ xantphos dichloride, 4,5-bis diphenylphosphino-9,9-dimethylxanthene dichloropalladium ii, 4,5-bis diphenylphosphino-9,9-dimethylxanthene palladium ii dichloride
PubChem CID 86280406
IUPAC Name (5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane;palladium(2+);dichloride
SMILES Cl[Pd++]Cl.CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 280°C to 287°C (decomposition)
Color Yellow
Quantity 1 g
Formula Weight 755.96
Concentration or Composition (by Analyte or Components) Pd 14.1%
Physical Form Crystalline Solid
Chemical Name or Material Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), Pd 12.1%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.