missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[bis(1,3-diphenylphosphino)propane]palladium(II), Thermo Scientific Chemicals

23.50 € - 62.11 €

Chemical Identifiers

CAS 59831-02-6
Molecular Formula C27H26Cl2P2Pd
Molecular Weight (g/mol) 589.77
MDL Number MFCD03844773
InChI Key LDFBXJODFADZBN-UHFFFAOYSA-L
Synonym [1,3-Bis(diphenylphosphino)propane]dichloropalladium(II)
PubChem CID 131664225
IUPAC Name 3-diphenylphosphanylpropyl(diphenyl)phosphane;palladium(2+);dihydrochloride
SMILES [Cl-].[Cl-].[Pd++].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15487625
SDS View Documents
Thermo Scientific Alfa Aesar
H26897.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11411450
SDS View Documents
Thermo Scientific Alfa Aesar
H26897.03
1 g
62.11 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

(1,3-Bis(diphenylphosphino)propane)palladium(II) chloride, used ubiquitously as a employed preparation of palladium for cross-coupling protocols in organic synthesis. It is also a useful organo-metallic chemical catalyst. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1,3-Bis(diphenylphosphino)propane)palladium(II) chloride, used ubiquitously as a employed preparation of palladium for cross-coupling protocols in organic synthesis. It is also a useful organo-metallic chemical catalyst. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in coupling reactions.

Solubility
Soluble in dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloro[bis(1,3-diphenylphosphino)propane]palladium(II), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.