Learn More
Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II), Pd 17.6%, Thermo Scientific Chemicals
Description
Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II) is used for the catalysis of styrene carbonylation, coupling of alkyl Grignard reagents with organic halides, selective monoalkylation of organic polyhalides, and modification of the dihalovinyl moiety of synthetic pyrethroids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 1 g |
Solubility Information | Soluble in ethanol,benzene,and ethereal solvents. |
Formula Weight | 603.8 |
Concentration or Composition (by Analyte or Components) | Pd 17.6% |
Physical Form | Lumps |
Chemical Name or Material | Dichloro[bis (1,4-diphenylphosphino)butane]palladium (II) |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Safety and Handling

- Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II)
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II)
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.