missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II), Pd 15%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Description
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) is a highly active, air-stable catalyst for Suzuki-Miyaura cross-coupling with aryl halides including 5- and 6-membered heteroaryl chlorides.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
CAS | 887919-35-9 |
Color | Yellow |
Molecular Formula | C32H56Cl2N2P2Pd |
MDL Number | MFCD09265123 |
Quantity | 1 g |
Synonym | bis di-tert-butyl 4-dimethylaminophenyl phosphine dichloropalladium ii, pd amphos cl2, pdcl2 amphos 2, a-taphos 2pdcl2, dichlorobis di-tert-butyl 4-dimethylaminophenyl phosphine palladium ii, alpha-taphos 2pdcl2, bis 4-di-tert-butylphosphino-n,n-dimethylbenzenamine palladium dichloride |
Solubility Information | Insoluble in water. |
InChI Key | DWOZNANUEDYIOF-UHFFFAOYSA-L |
SMILES | Cl[Pd]Cl.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C |
IUPAC Name | 4-ditert-butylphosphanyl-N,N-dimethylaniline;dichloropalladium |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'