Learn More
Dichlorodimethylsilane, 98+%, Thermo Scientific Chemicals
44.52 € - 102.51 €
Identifiants chimiques
CAS | 75-78-5 |
---|---|
Molecular Formula | C2H6Cl2Si |
Molecular Weight (g/mol) | 129.055 |
MDL Number | MFCD00000491 |
InChI Key | LIKFHECYJZWXFJ-UHFFFAOYSA-N |
Synonym | dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane |
PubChem CID | 6398 |
IUPAC Name | dichloro(dimethyl)silane |
SMILES | C[Si](C)(Cl)Cl |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
11338635
|
Thermo Scientific Alfa Aesar
L12133.22 |
100 g |
44.52 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348635
|
Thermo Scientific Alfa Aesar
L12133.36 |
500 g |
64.22 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358635
|
Thermo Scientific Alfa Aesar
L12133.0B |
1000 g |
102.51 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dichlorodimethylsilane is used to prepare a resin bound siloxane with tertiary alcohols and it is also used as a reagent for synthesis of optically active ansa-mettallocene polymerization catalysts. It acts as a precursor to silicone and polysilane compounds. It is also used in the glass coating to protect it from micro particles. It is involved in the preparation of resin bound siloxane with reactivity towards tertiary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDichlorodimethylsilane is used to prepare a resin bound siloxane with tertiary alcohols and it is also used as a reagent for synthesis of optically active ansa-mettallocene polymerization catalysts. It acts as a precursor to silicone and polysilane compounds. It is also used in the glass coating to protect it from micro particles. It is involved in the preparation of resin bound siloxane with reactivity towards tertiary alcohols.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with alcohols, amines and strong bases.
Identifiants chimiques
75-78-5 | |
129.055 | |
LIKFHECYJZWXFJ-UHFFFAOYSA-N | |
6398 | |
C[Si](C)(Cl)Cl |
C2H6Cl2Si | |
MFCD00000491 | |
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane | |
dichloro(dimethyl)silane |
Spécification
70°C to 71°C | |
75-78-5 | |
(CH3)2SiCl2 | |
UN1162 | |
−5°C (23°F) | |
Immiscible with water. | |
C[Si](C)(Cl)Cl | |
129.055 | |
129.06 | |
1.4045 | |
1.068 |
∼-76°C | |
C2H6Cl2Si | |
MFCD00000491 | |
605287 | |
dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane | |
LIKFHECYJZWXFJ-UHFFFAOYSA-N | |
dichloro(dimethyl)silane | |
6398 | |
99% | |
Moisture sensitive | |
Dichlorodimethylsilane |
Sécurité et traitement
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H331-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLDICHLOROSILANE
missing translation for 'einecsNumber' : 200-901-0
missing translation for 'rtecsNumber' : VV3150000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only