missing translation for 'onlineSavingsMsg'
Learn More
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10387841

Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10387841
1 L, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10387841

Brand: Thermo Scientific Acros 348460010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Dichloromethane, 99.8%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine, 1L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless, >120 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2, 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mmShow MoreShow Less
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Decomposition Information >120°C
Melting Point -97.0°C
Color Colorless
Density 1.3250g/mL
Boiling Point 39.0°C to 40.0°C
Infrared Spectrum Authentic
Assay Percent Range 99.8%
Packaging AcroSeal™ Glass Bottle
Linear Formula CH2Cl2
Quantity 1 L
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Stabilizer 50ppm (approx.) amylene
Formula Weight 84.93
Vapor Pressure 475hPa at 20°C
Percent Purity 99.8%
Grade Extra Dry over Molecular Sieve
Physical Form Liquid
Chemical Name or Material Dichloromethane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2495453 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number348461000,348460025,348465000,348460010
Lot Number2524002 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number348461000,348460025,348465000,348460010
Lot Number2402361 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number348461000,348460025,348465000,348460010
Lot Number2412973 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number348461000,348460025,348465000,348460010
Lot Number2416196 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number348461000,348460025,348465000,348460010
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Dichloromethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.