missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, 99.8%, for HPLC, stabilized with amylene, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC

Specifications
Specifications
Melting Point | -97.0°C |
Absorbance | (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 260nm, 0.005 max. at 270nm, 0.005 max. at 280nm, 0.02 max. at 250nm, 0.2 max. at 240nm, 1 max. at 233nm |
Acidity | 0.0001 meq/g max. |
Density | 1.3250g/mL |
Boiling Point | 39.0°C to 40.0°C |
Quantity | 1 L |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | 99.8% |
Residue after Evaporation | 0.0002% max. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1738782 | Certificate of Analysis | 28/01/2025 | 268330025,268330010 |
1875662 | Certificate of Analysis | 28/01/2025 | 268330025,268330010 |
2058988 | Certificate of Analysis | 28/01/2025 | 268330025,268330010 |
1736629 | Certificate of Analysis | 27/01/2025 | 268330025,268330010 |
1738523 | Certificate of Analysis | 27/01/2025 | 268330025,268330010 |
Safety and Handling


Product Identifier
- Dichloromethane
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H351-Suspected of causing cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dichloromethane, 99.8%, for HPLC, stabilized with amylene, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction