missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, 99.8%, for HPLC, stabilized with methanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for HPLC

Specifications
Specifications
Melting Point | -97.0°C |
Absorbance | (1 cm cell vs HPLC−grade water), 0.005 max. at 260nm, 0.005 max. at 270nm, 0.005 max. at 280nm, 0.02 max. at 250nm, 0.2 max. at 240nm, 1 max. at 233nm |
Acidity | 0.0001 meq/g max. |
Density | 1.3250g/mL |
Boiling Point | 39.0°C to 40.0°C |
Quantity | 2.5 L |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | 99.80% |
Residue after Evaporation | 0.0002% max. |
Show More |
Safety and Handling


Product Identifier
- Dichloromethane, stabilized with methanol
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dichloromethane, 99.8%, for HPLC, stabilized with methanol, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction