missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane ≥99.8% stabilised, ULTRA RESI-ANALYZED™ J.T. Baker™

Product Code. 10709131
Click to view available options
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

For organic residue analysis

  • Stabilised with a methanol-alkene preservative system
  • Packaged under inert gas
  • For Laboratory, Research, or Manufacturing Use

Specifications

Melting Point -95°C
CAS 75-09-2
Color ≤10
Density 1.322 g/cm³ (20°C)
Boiling Point 39.8°C (1013 hPa)
Quantity 2.5 L
Packaging Glass Bottle
Assay Percent Range ≥99.8 %
Residue after Evaporation ≤1.0 ppm
Molecular Formula CH2Cl2
Titratable Acid ≤0.3 μeq/g
MDL Number MFCD00000881
Water ≤0.02%
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P281-Use personal protective equipment as required.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.