missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Laboratory Reagent, ≥99.9% (without stabilizer, GC), contains 0.1-0.4% ethanol as stabilizer, Honeywell™
Click to view available options
Quantity:
4 x 5 L
Packaging:
Aluminum Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
CAS | 75-09-2 |
Molecular Formula | CH2Cl2 |
MDL Number | MFCD00000881 |
Beilstein | 1730800 |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
SMILES | ClCCl |
IUPAC Name | dichloromethane |
Molecular Weight (g/mol) | 84.93 |
PubChem CID | 6344 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'