Learn More
Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
41.61 € - 492.45 €
Chemical Identifiers
CAS | 75-09-2, 64-17-5 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mmShow More |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10103510
|
Thermo Scientific Acros
124050010 |
1 L | Glass Bottle |
41.61 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10011180
|
Thermo Scientific Acros
124050025 |
2.5 L | Glass Bottle |
85.53 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15616845
|
Thermo Scientific Acros
124050050 |
5 L | Plastic Bottle |
147.74 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10477510
|
Thermo Scientific Acros
124050250 |
25 L | Metal Drum |
492.45 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-09-2, 64-17-5 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mmShow More | |
CHEBI:15767 | |
ClCCl |
Specifications
-97.0°C | |
1.3250g/mL | |
Liquid | |
99+% | |
1.325 | |
CH2Cl2 | |
0.43 mPa.s (20°C) | |
0.1% max. (K.F.) | |
15, 6135 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
≥99% | |
Dichloromethane |
75-09-2, 64-17-5 | |
39.0°C to 40.0°C | |
1.4230 to 1.4250 | |
0.05% max. | |
CH2Cl2 | |
MFCD00000881 | |
01, 60 | |
01,676; 02,273; 04,337; 07,239; 13,106 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.93 | |
Extra Pure |
Safety and Handling
GHS P Statement
Suspected of causing cancer.
GHS P Statement
Use personal protective equipment as required. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Dichloromethane, stabilized with ethanol
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Methylene chloride,Ethyl alcohol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.