missing translation for 'onlineSavingsMsg'
Learn More
Dichloromethane, 99+%, stab. with ca. 50ppm amylene, Thermo Scientific Chemicals Product Code.: 11367876

Dichloromethane, 99+%, stab. with ca. 50ppm amylene, Thermo Scientific Chemicals

Product Code. 11367876
500 mL
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11367876

Brand: Thermo Scientific Alfa Aesar L13089.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

Solubility
Miscible with ethyl acetate, alcohol, hexanes, methanol, diethyl ether, n-octanol, acetone benzene, carbon tetrachloride, diethyl ether and chloroform. Immiscible with water.

Notes
Keep the container tightly closed in a cool, well-ventilated area. Incompatible with alkali metals, aluminum, strong oxidizing agents, bases, amines, magnesium, strong acids strong bases and vinyl compounds.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -95°C
Density 1.325
Boiling Point 39°C to 40°C
Quantity 500 mL
Odor Sweet
Refractive Index 1.4244
UN Number UN1593
Beilstein 1730800
Merck Index 14,6063
Solubility Information Miscible with ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. Immiscible with water.
Formula Weight 84.93
Percent Purity ≥99%
Chemical Name or Material Dichloromethane, stabilized with ca. 50ppm amylene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2411744 Certificate TypeCertificate of Analysis Date04/10/2024 Catalog NumberL13089.0F,L13089.AP,L13089.AU
Lot Number2493927 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL13089.0F,L13089.AP,L13089.AU
Lot Number10250085 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberL13089.0F,L13089.AP,L13089.AU
Lot Number10250086 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog NumberL13089.0F,L13089.AP,L13089.AU
Lot Number10239056 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL13089.0F,L13089.AP,L13089.AU

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloromethane, 99+%, stab. with ca. 50ppm amylene, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.