missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane 99+%, stabilized with amylene, pure, Thermo Scientific Chemicals
41.00 € - 411.00 €
Chemical Identifiers
CAS | 75-09-2, 513-35-9 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13168573
|
Thermo Scientific Acros
383780010 |
1 L |
41.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10590873
|
Thermo Scientific Acros
383780025 |
2.5 L |
84.20 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10046082
|
Thermo Scientific Acros
383780050 |
5 L | |||||||
10650735
|
Thermo Scientific Acros
383780250 |
25 L |
411.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-09-2, 513-35-9 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
-97.0°C | |
1.3250g/mL | |
Glass Bottle | |
1.325 | |
CH2Cl2 | |
0.43 mPa.s (20°C) | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.93 | |
Extra Pure |
75-09-2, 513-35-9 | |
39.0°C to 40.0°C | |
99+% | |
CH2Cl2 | |
MFCD00000881 | |
01,676; 02,273; 04,337; 07,239; 13,106 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
99+% | |
Dichloromethane |
Safety and Handling
missing translation for 'einecsNumber' : 200-838-9
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Dichloromethane
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H351-Suspected of causing cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only