missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, ACS Reagent, contains 40-150 amylene as stabilizer, ≥99.5%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
Melting Point | -97°C (literature) |
CAS Min % | ≥99.0000% |
CAS Max % | ≥99.0000% |
Density | 1.325 g/mL at 25 °C(lit.) |
Boiling Point | 39°C to 41°C |
Quantity | 10 L |
Packaging | Stainless Steel Can |
Linear Formula | CH2Cl2 |
UN Number | UN1593 |
Beilstein | 1730800 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'