missing translation for 'onlineSavingsMsg'
Learn More
Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, stab. with amylene, Thermo Scientific Chemicals Product Code.: 11354797

Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, stab. with amylene, Thermo Scientific Chemicals

Product Code. 11354797
250 mL
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11354797

Brand: Thermo Scientific Alfa Aesar

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solvent For Anhydrous Reactions, Water <0.01%

Solvent for anhydrous reactions, water <0.01%.Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point - 95°C
Color Colorless
Density 1.325 g/cm3 at 20°C
Boiling Point 39°C to 40°C
Ignition Point 605°C (1121°F)
Assay Percent Range >99.7%
Odor Sweet
Refractive Index 1.4244
Quantity 250 mL
Beilstein 1730800
Solubility Information Miscible with ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. Immiscible with water.
Formula Weight 84.93
Vapor Pressure 475 hPa (356mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Dichloromethane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW30K950 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog Number041835.AK,041835.K2,041835.M1
Lot NumberX04K968 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number041835.AK,041835.K2,041835.M1
Lot NumberW30K951 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog Number041835.AK,041835.K2,041835.M1
Lot NumberW16K898 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number041835.AK,041835.K2,041835.M1
Lot NumberW16K897 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number041835.AK,041835.K2,041835.M1
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : dichloromethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable ChemSeal™ bottles, stab. with amylene, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.