missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, For pesticide residue analysis, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Specifications
Melting Point | -97°C (literature) |
CAS Min % | ≥99.0000% |
CAS Max % | ≥99.0000% |
Density | 1.325 g/mL at 25 °C(lit.) |
Boiling Point | 39°C to 41°C |
Quantity | 4 x 2.5 L |
Packaging | Glass Bottle |
Linear Formula | CH2Cl2 |
UN Number | UN1593 |
Beilstein | 1730800 |
Show More |
Safety and Handling
Product Identifier
- Dichloromethane
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'