missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Primar™, for Trace Metal Analysis, Stabilised with Amylene, Fisher Chemical
Click to view available options
2.5L, Amber glass bottle
Specifications
Specifications
Melting Point | -97°C |
CAS | 75-09-2 |
Color | Colorless |
Boiling Point | 39°C |
Quantity | 2.5 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Molecular Formula | CH2Cl2 |
MDL Number | MFCD00000881 |
Vapor Pressure | 350mbar at 20°C |
Show More |
Safety and Handling
Product Identifier
- Dichloromethane
- Warning
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H351-Suspected of causing cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'