missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Puriss. p.a., ACS Reagent, Reag. ISO, ≥99.9% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Spezifikation
Melting Point | -97°C |
Color | Colorless |
Appearance | Liquid |
Density | 1.325g/mL at 25°C, 1.320 to 1.330g/Lat 20°C |
Boiling Point | 39.8°C to 40°C |
Quantity | 4 x 2.5 L |
Explosion Limit | 0.22 |
Ignition Point | 1223°F |
Packaging | Glass Bottle |
Refractive Index | n20/D 1.4235 to 1.4245 |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Dichloromethane
- Warning
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : dichloromethane
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'