missing translation for 'onlineSavingsMsg'
Learn More
Dichloromethane 99.8+%, for Residue Analysis, Distol™, Stabilised with Amylene, Fisher Chemical Product Code.: 10132140

Dichloromethane 99.8+%, for Residue Analysis, Distol™, Stabilised with Amylene, Fisher Chemical

Product Code. 10132140
1L, Amber glass bottle
Click to view available options
1L, Amber glass bottle
This item is not returnable. View return policy

Product Code. 10132140

Brand: Fisher Chemical D/1853/15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point -97°C
CAS 75-09-2
Color Colorless
Boiling Point 39°C
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Molecular Formula CH2Cl2
MDL Number MFCD00000881
Vapor Pressure 350mbar at 20°C
Viscosity 0.43 mPaS at 20°C
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
PubChem CID 6344
ChEBI CHEBI:15767
Formula Weight 84.93g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloromethane 99.8+%, for Residue Analysis, Distol™, Stabilised with Amylene, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.