missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals

21.21 € - 117.59 €

Chemical Identifiers

CAS 124-70-9
Molecular Formula C3H6Cl2Si
Molecular Weight (g/mol) 141.066
MDL Number MFCD00000492
InChI Key YLJJAVFOBDSYAN-UHFFFAOYSA-N
Synonym dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane
PubChem CID 31299
IUPAC Name dichloro-ethenyl-methylsilane
SMILES C[Si](C=C)(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394509
View Documents
Thermo Scientific Alfa Aesar
B23119.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11436336
View Documents
Thermo Scientific Alfa Aesar
B23119.22
100 g
34.07 €
100g
Please to purchase this item. Need a web account? Register with us today!
11446336
View Documents
Thermo Scientific Alfa Aesar
B23119.36
500 g
117.59 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124-70-9
141.066
YLJJAVFOBDSYAN-UHFFFAOYSA-N
31299
C[Si](C=C)(Cl)Cl
C3H6Cl2Si
MFCD00000492
dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane
dichloro-ethenyl-methylsilane

Specifications

92°C to 93°C
124-70-9
MFCD00000492
1740822
dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane
C[Si](C=C)(Cl)Cl
141.066
141.07
Acrid
Moisture sensitive
Dichloromethylvinylsilane
-78°C
C3H6Cl2Si
UN2985
4°C (39°F)
YLJJAVFOBDSYAN-UHFFFAOYSA-N
dichloro-ethenyl-methylsilane
31299
97%
1.43
1.087
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 204-710-3

missing translation for 'rtecsNumber' : VV3533000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.