missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophenylsilane, 96%, Thermo Scientific Chemicals

Product Code. 11356916
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from moisture, oxidizing agents. Incompatible materials are bases, moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1631-84-1
Molecular Formula C6H5Cl2Si
Molecular Weight (g/mol) 176.09
MDL Number MFCD00039296
InChI Key XNAFLNBULDHNJS-UHFFFAOYSA-N
Synonym phenyldichlorosilane, dichlorophenylsilane, dichloro phenyl silane, dichloro phenyl silicon, benzene, dichlorosilyl, phenyl dichlorosilane, phenyl hydrogen dichlorosilane, phenyl-dichlorosilane, phenyldichloro silane, dichloro phenyl silyl
PubChem CID 6327324
SMILES Cl[Si](Cl)C1=CC=CC=C1

Specifications

Boiling Point 181°C
Quantity 5 g
UN Number UN2986
Beilstein 2433865
Flash Point 48°C (118°F)
Solubility Information Hydrolyzes in water.
IUPAC Name dichloro(phenyl)silicon
Formula Weight 177.11
Percent Purity 96%
Refractive Index 1.523
Sensitivity Moisture sensitive
Density 1.212
Chemical Name or Material Dichlorophenylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.