Learn More
Diclofenac diethylamine, MedChemExpress
MedChemExpress Diclofenac diethylamine is a potent and nonselective anti-inflammatory agent, acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. Diclofenac diethylamine induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.
Brand:  Medchem Express HY-15036A
Encompass_Preferred
 View more versions of this product
Product Code. 16448887
  • 90.45 € / 5g
Explore more special offers
Description
Description
Diclofenac diethylamine is a potent and nonselective anti-inflammatory agent, acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. Diclofenac diethylamine induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.
Specifications
Specifications
Diclofenac diethylamine
Off-White
C18H22Cl2N2O2
DMSO : 150 mg/mL (406.18 mM; Need ultrasonic)
369.29
99.55%
Solid
78213-16-8
Research
5 g
O=C(O)CC1=CC=CC=C1NC2=C(Cl)C=CC=C2Cl.CCNCC
369.29
Research
COVID-19-immunoregulation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Diclofenac diethylamine, MedChemExpress