missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicumyl peroxide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Dicumyl peroxide (DCP) is used:
- in vulcanization of rubber.
- as a cross linking agent in the synthesis of polylactic acid composite fibers.
- in the preparation of polyethylene composites.
- in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dicumyl peroxide (DCP) is used: • in vulcanization of rubber. • as a cross linking agent in the synthesis of polylactic acid composite fibers. • in the preparation of polyethylene composites. • in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.
Solubility
Soluble in chloroform and most organic solvents. Insoluble in water.
Notes
Store at 4°C. Incompatible with oxidizing agents and acids.
Dicumyl peroxide (DCP) is used: • in vulcanization of rubber. • as a cross linking agent in the synthesis of polylactic acid composite fibers. • in the preparation of polyethylene composites. • in the synthesis of polyamide 112/ethylene vinyl acetate copolymer blends.
Solubility
Soluble in chloroform and most organic solvents. Insoluble in water.
Notes
Store at 4°C. Incompatible with oxidizing agents and acids.
Specifications
Specifications
Melting Point | 39°C to 41°C |
Density | 1.56 |
Linear Formula | [C6H5C(CH3)2]2O2 |
Quantity | 100 g |
UN Number | UN3110 |
Beilstein | 2056090 |
Solubility Information | Soluble in chloroform and most organic solvents. Insoluble in water. |
Formula Weight | 270.37 |
Percent Purity | 98% |
Chemical Name or Material | Dicumyl peroxide |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1A
- Skin corrosion/irritation Category 2
- H242-Heating may cause a fire.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H360D-May damage the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction