missing translation for 'onlineSavingsMsg'
Learn More

Dicyandiamide, 99.5%, Thermo Scientific Chemicals

Product Code. 10242462
Click to view available options
Quantity:
5 g
25 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-58-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008066
InChI Key QGBSISYHAICWAH-UHFFFAOYSA-N
Synonym dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
PubChem CID 10005
SMILES NC(N)=NC#N

Specifications

Melting Point 208.0°C to 211.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Glass Bottle
Quantity 5 g
Fieser 01,229
Merck Index 15, 3103
Solubility Information Solubility in water: 32g/L (20°C). Other solubilities: 38g/L in methanol (20°C)
IUPAC Name 2-cyanoguanidine
Formula Weight 84.08
Percent Purity 99.5%
Physical Form Crystalline Powder
Chemical Name or Material Dicyandiamide
Show More Show Less
Product Identifier
  • Dicyandiamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'