missing translation for 'onlineSavingsMsg'
Learn More
Dicyandiamide, 99%, Thermo Scientific Chemicals Product Code.: 11384509

Dicyandiamide, 99%, Thermo Scientific Chemicals

Product Code. 11384509
250 g
Click to view available options
Quantity:
250 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11384509

Brand: Thermo Scientific Alfa Aesar A10451.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyandiamide is used in the synthesis of barbiturates. It is used as a stabilizer of ammonium dinitramide melt. It is used as hardener.

Solubility
Soluble in Water (32g/L ) at 20°C

Notes
Store at 2-8°C. Incompatible with strong acids, strong oxidizing agents, strong acids, strong bases, ammonium nitrate, potassium chlorates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-58-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008066
InChI Key QGBSISYHAICWAH-UHFFFAOYSA-N
Synonym dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
PubChem CID 10005
IUPAC Name 2-cyanoguanidine
SMILES NC(N)=NC#N

Specifications

Melting Point 208°C to 211°C
Density 1.4
Assay Percent Range 99%
Odor Odorless
Quantity 250 g
Beilstein 605637
Merck Index 14,3092
Solubility Information Soluble in Water (32g/L ) at 20°C
Formula Weight 84.08
Percent Purity 99%
Chemical Name or Material Dicyandiamide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dicyandiamide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.